missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| L-(-)-Glucose | |
| White | |
| MFCD00148913 | |
| l-glucose, l---glucose, levoglucose, l--glucose, 2s,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-glucose, aldehydo-l-gluco-hexose, d-glucose-13c6,d7, levoglucose usan, glucose, l-isomer | |
| C(C(C(C(C(C=O)O)O)O)O)O | |
| 180.156 | |
| CHEBI:37626 | |
| ≥98.0% (GC) |
| 921-60-8 | |
| C6H12O6 | |
| 5 g | |
| GZCGUPFRVQAUEE-VANKVMQKSA-N | |
| (2S,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal | |
| 10954115 | |
| 180.16 | |
| Crystalline Powder |
Chemical Identifiers
| 921-60-8 | |
| 180.156 | |
| GZCGUPFRVQAUEE-VANKVMQKSA-N | |
| 10954115 | |
| (2S,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal |
| C6H12O6 | |
| MFCD00148913 | |
| l-glucose, l---glucose, levoglucose, l--glucose, 2s,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-glucose, aldehydo-l-gluco-hexose, d-glucose-13c6,d7, levoglucose usan, glucose, l-isomer | |
| CHEBI:37626 | |
| C(C(C(C(C(C=O)O)O)O)O)O |
Safety and Handling
RTECSNumber : LZ6610000
TSCA : No