missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Glucose, anhydrous, 98%, Thermo Scientific Chemicals

$223.44

Chemical Identifiers

CAS 921-60-8
Molecular Formula C6H12O6
MDL Number MFCD00148913
PubChem CID 10954115
ChEBI CHEBI:37626
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1749603
View Documents Promotion Details
N/A 1 g
Each for $223.44
Only null left
 
Description

Description

L-(-)-Glucose is an enantiomer of the more common D-glucose a naturally occurring carbohydrate used in an abundance of cellular processes. L-Glucose is a synthetic sugar used in the formation of L-Glucose Pen taacetate, a potential therapeutic agent regarding type II diabetes. In addition, L-glucose can be used as a colon cleansing agent for before a colonoscopy procedure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-Glucose is an enantiomer of the more common D-glucose a naturally occurring carbohydrate used in an abundance of cellular processes. L-Glucose is a synthetic sugar used in the formation of L-Glucose Pen taacetate, a potential therapeutic agent regarding type II diabetes. In addition, L-glucose can be used as a colon cleansing agent for before a colonoscopy procedure.

Solubility
Soluble in water, alcohol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

921-60-8
MFCD00148913
CHEBI:37626
C6H12O6
10954115

Specifications

921-60-8
C6H12O6
1724626
Soluble in water,alcohol.
−53° (c=10 in Water, 24h)
(2S,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
10954115
180.16
L-(-)-Glucose, anhydrous
148°C to 157°C
MFCD00148913
l-glucose, l---glucose, levoglucose, l--glucose, 2s,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-glucose, aldehydo-l-gluco-hexose, d-glucose-13c6,d7, levoglucose usan, glucose, l-isomer
GZCGUPFRVQAUEE-VANKVMQKSA-N
C(C(C(C(C(C=O)O)O)O)O)O
180.156
CHEBI:37626
98%
Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 213-068-3

RTECSNumber : LZ6610000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only