missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Glucose, 98%

Catalog No. ac241920010
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC241920010 1 g Glass bottle
AC241922500 250 mg Glass bottle
AC241920050 5 g Glass bottle
AC241920250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC241920010 Supplier Thermo Scientific Chemicals Supplier No. 241920010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 921-60-8 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 921-60-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00148913
InChI Key GZCGUPFRVQAUEE-VANKVMQKSA-N
Synonym l-glucose, l---glucose, levoglucose, l--glucose, 2s,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-glucose, aldehydo-l-gluco-hexose, d-glucose-13c6,d7, levoglucose usan, glucose, l-isomer
PubChem CID 10954115
ChEBI CHEBI:37626
IUPAC Name (2S,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 153.0°C to 156.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition − 52.00 (20.00°C c=10,H2O,NH3)
Specific Rotation − 52.00
Solubility Information (10% in water) Clear colorless
Formula Weight 180.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L(-)-Glucose
Show More Show Less
EINECSNumber 213-068-3

RUO – Research Use Only