missing translation for 'onlineSavingsMsg'
Learn More

L-gamma-Glutamyl-p-nitroanilide Monohydrate 98.0+%, TCI America™

Catalog No. G00655G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G00655G 5 g
G00651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G00655G Supplier TCI America Supplier No. G00655G

Chemical Identifiers

CAS 7300-59-6
Molecular Formula C11H13N3O5
Molecular Weight (g/mol) 267.24
MDL Number MFCD00036218
InChI Key WMZTYIRRBCGARG-VIFPVBQESA-N
Synonym s-2-amino-5-4-nitrophenyl amino-5-oxopentanoic acid, l-glutamine, n-4-nitrophenyl, n-4-nitrophenyl-l-glutamine, l-gamma-glutamyl-p-nitroanilide, unii-81sz84nxws, l-2-amino-4'-nitroglutaranilic acid, gamma-glutamine-4-nitroanilide, gamma-l-glutamyl-4-nitroanilide, 81sz84nxws, chembl6251
PubChem CID 81732
IUPAC Name (2S)-2-azaniumyl-4-[(4-nitrophenyl)carbamoyl]butanoate
SMILES [NH3+][C@@H](CCC(=O)NC1=CC=C(C=C1)[N+]([O-])=O)C([O-])=O

Specifications

Melting Point 178°C
Color Yellow
Quantity 5 g
Formula Weight 267.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-gamma-Glutamyl-p-nitroanilide Monohydrate
EINECSNumber (9)-2059
TSCA Yes