missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Galactose 98.0+%, TCI America™

Catalog No. p-7130216
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0267100MG 100 mg
G0267500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0267100MG Supplier TCI America Supplier No. G0267100MG
Only null left

Chemical Identifiers

CAS 15572-79-9
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00063833
InChI Key GZCGUPFRVQAUEE-DPYQTVNSSA-N
Synonym aldehydo-l-galactose, 2s,3r,4r,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-galacto-hexose, unii-s93uii1dw8, s93uii1dw8, galactose, l
PubChem CID 84996
ChEBI CHEBI:37617
IUPAC Name (2S,3R,4R,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Color White
Quantity 100 mg
Formula Weight 180.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Galactose