missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(-)-Galactose, 98%

Catalog No. AAB21448MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB21448MC 100 mg
AAB21448ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB21448MC Supplier Thermo Scientific Chemicals Supplier No. B21448MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15572-79-9 | C6H12O6 | 180.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15572-79-9
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00063833
InChI Key GZCGUPFRVQAUEE-DPYQTVNSSA-N
Synonym aldehydo-l-galactose, 2s,3r,4r,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-galacto-hexose, unii-s93uii1dw8, s93uii1dw8, galactose, l
PubChem CID 84996
ChEBI CHEBI:37617
IUPAC Name (2S,3R,4R,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 163°C to 165°C
Quantity 100 mg
Beilstein 1724622
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material L-(-)-Galactose
EINECSNumber 239-630-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only