missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(-)-Fucose, 98%

Catalog No. p-7023346
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1678903 1 g
AAA1678906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1678903 Supplier Thermo Scientific Chemicals Supplier No. A1678903

CAS: 2438-80-4 | C6H12O5 | 164.16 g/mol

L-(-)-Fucose finds application in cosmetics, pharmaceuticals and dietary supplements. It is used in the determination of antigen in A and B blood group. It is also used in the selection-mediated leukocyte-endothelial adhesion and host-microbe interactions. It is also used in anti aging creams as well as to promote the accelerated healing of wounds and to reduce allergy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-Fucose finds application in cosmetics, pharmaceuticals and dietary supplements. It is used in the determination of antigen in A and B blood group. It is also used in the selection-mediated leukocyte-endothelial adhesion and host-microbe interactions. It is also used in anti aging creams as well as to promote the accelerated healing of wounds and to reduce allergy.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.

Chemical Identifiers

CAS 2438-80-4
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00135607
InChI Key SHZGCJCMOBCMKK-DHVFOXMCSA-N
Synonym l---fucose, aldehydo-l-fucose, l-galactose, 6-deoxy, 2s,3r,4r,5s-2,3,4,5-tetrahydroxyhexanal, unii-28ryy2iv3f, l---rhodeose, 28ryy2iv3f, l--fucose, l-rhodeose, 3h-fucose
PubChem CID 3034656
ChEBI CHEBI:48204
SMILES C[C@@H]1OC(O)[C@@H](O)[C@H](O)[C@@H]1O

Specifications

Melting Point 139°C to 142°C
Quantity 1 g
Beilstein 1422661
Sensitivity Hygroscopic
Merck Index 14,4279
Solubility Information Soluble in water.
Optical Rotation −75° (c=10 in Water)
IUPAC Name (2S,3R,4R,5S)-2,3,4,5-tetrahydroxyhexanal
Formula Weight 164.16
Percent Purity 98%
Chemical Name or Material L-(-)-Fucose
Show More Show Less
EINECSNumber 219-452-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only