missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Fucose, 97%

Catalog No. AC225880010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC225880010 1 g Glass bottle
AC225880050 5 g Glass bottle
AC225880250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC225880010 Supplier Thermo Scientific Chemicals Supplier No. 225880010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2438-80-4 | C6H12O5 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2438-80-4
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00135607
InChI Key SHZGCJCMOBCMKK-DHVFOXMCSA-N
Synonym l---fucose, aldehydo-l-fucose, l-galactose, 6-deoxy, 2s,3r,4r,5s-2,3,4,5-tetrahydroxyhexanal, unii-28ryy2iv3f, l---rhodeose, 28ryy2iv3f, l--fucose, l-rhodeose, 3h-fucose
PubChem CID 3034656
ChEBI CHEBI:48204
SMILES C[C@@H]1OC(O)[C@@H](O)[C@H](O)[C@@H]1O

Specifications

Melting Point 128.0°C to 141.0°C
Color Beige to Gray-Purple
Loss on Drying 0.5% max. (60°C, 4 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 31, 78
Merck Index 15, 4308
Specific Rotation Condition - 78.00 (20.00°C c=4,H2O,NH3)
Specific Rotation − 78.00
Solubility Information Solubility in water: soluble in water
IUPAC Name (3S,4R,5S,6S)-6-methyloxane-2,3,4,5-tetrol
Formula Weight 164.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material L(-)-Fucose, 97%
Show More Show Less
EINECSNumber 219-452-7
TSCA TSCA

RUO – Research Use Only