missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Fructose 95.0+%, TCI America™

Catalog No. p-7129610
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0317100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0317100MG Supplier TCI America Supplier No. F0317100MG
Only null left

Chemical Identifiers

CAS 7776-48-9
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD05662378
InChI Key BJHIKXHVCXFQLS-FUTKDDECSA-N
PubChem CID 5460024
ChEBI CHEBI:37724
IUPAC Name (3R,4S,5S)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Color White
Quantity 100 mg
Formula Weight 180.16
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material L-(+)-Fructose