accessibility menu, dialog, popup

UserName

OH(S)OHOHO(R)OH(R)OH

CAS RN 7776-48-9

IUPAC Name: (3R,4R,5S)-2,5-bis(hydroxymethyl)oxolane-2,3,4-triol
Synonyms: L-fructose L-fru L-arabino-hexulose
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: RFSUNEUAIZKAJO-NSHGFSBMSA-N
SMILES: OC[C@@H]1OC(O)(CO)[C@H](O)[C@H]1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results