missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014669
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014669 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014669 Supplier Merck Emd Millipore Supplier No. PHR1323500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L-Cysteine a sulfur-containing, proteingenic non-essential amino acid useful in numerous biological applications.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 56-89-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Linear Formula [-SCH2 CH(NH2)CO2 H]2
MDL Number MFCD00064228
Melting Point >240°C
Synonym L-Cystine; (R,R)-3,3′-Dithiobis(2-aminopropionicacid)
Type Certified Reference Material
Quantity 500 mg
Formula Weight 240.30
Grade Certified Reference Material