missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine disodium salt monohydrate, 98+%

CAS: 199329-53-8 | C6H12N2Na2O5S2 | 302.271 g/mol

$171.95 - $600.21

Chemical Identifiers

CAS 199329-53-8
Molecular Formula C6H12N2Na2O5S2
Molecular Weight (g/mol) 302.271
MDL Number MFCD00243291
InChI Key TVPDKJXPEMKMDM-BLZBDRIDSA-L
Synonym disodium l-cystine anion hydrate, l-cystine disodium salt monohydrate, h-cys-oh 2.2na.h2o, c6h10n2o4s2.2na.h2o
PubChem CID 71299670
IUPAC Name disodium;(2R)-2-amino-3-[[(2R)-2-amino-2-carboxylatoethyl]disulfanyl]propanoate;hydrate
SMILES C(C(C(=O)[O-])N)SSCC(C(=O)[O-])N.O.[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6307118
View Documents
Thermo Scientific Chemicals
J6307118
50 g
Each for $171.95
Only null left
 
AAJ6307130
View Documents
Thermo Scientific Chemicals
J6307130
250 g
Each for $600.21
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

199329-53-8
302.271
TVPDKJXPEMKMDM-BLZBDRIDSA-L
71299670
C(C(C(=O)[O-])N)SSCC(C(=O)[O-])N.O.[Na+].[Na+]
C6H12N2Na2O5S2
MFCD00243291
disodium l-cystine anion hydrate, l-cystine disodium salt monohydrate, h-cys-oh 2.2na.h2o, c6h10n2o4s2.2na.h2o
disodium;(2R)-2-amino-3-[[(2R)-2-amino-2-carboxylatoethyl]disulfanyl]propanoate;hydrate

Specifications

199329-53-8
C6H12N2Na2O5S2
50 g
TVPDKJXPEMKMDM-BLZBDRIDSA-L
disodium;(2R)-2-amino-3-[[(2R)-2-amino-2-carboxylatoethyl]disulfanyl]propanoate;hydrate
71299670
≥98%
L-Cystine disodium salt monohydrate
White
MFCD00243291
disodium l-cystine anion hydrate, l-cystine disodium salt monohydrate, h-cys-oh 2.2na.h2o, c6h10n2o4s2.2na.h2o
C(C(C(=O)[O-])N)SSCC(C(=O)[O-])N.O.[Na+].[Na+]
302.271
302.27 (284.26 Anhydrous)
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 265-025-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only