Learn More
L-Cystine disodium salt monohydrate, 98+%
CAS: 199329-53-8 | C6H12N2Na2O5S2 | 302.271 g/mol
Supplier: Thermo Scientific Chemicals J6307130
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| L-Cystine disodium salt monohydrate | |
| White | |
| MFCD00243291 | |
| disodium l-cystine anion hydrate, l-cystine disodium salt monohydrate, h-cys-oh 2.2na.h2o, c6h10n2o4s2.2na.h2o | |
| C(C(C(=O)[O-])N)SSCC(C(=O)[O-])N.O.[Na+].[Na+] | |
| 302.271 | |
| 302.27 (284.26 Anhydrous) | |
| Powder |
| 199329-53-8 | |
| C6H12N2Na2O5S2 | |
| 250 g | |
| TVPDKJXPEMKMDM-BLZBDRIDSA-L | |
| disodium;(2R)-2-amino-3-[[(2R)-2-amino-2-carboxylatoethyl]disulfanyl]propanoate;hydrate | |
| 71299670 | |
| ≥98% |
Chemical Identifiers
| 199329-53-8 | |
| 302.271 | |
| TVPDKJXPEMKMDM-BLZBDRIDSA-L | |
| 71299670 | |
| C(C(C(=O)[O-])N)SSCC(C(=O)[O-])N.O.[Na+].[Na+] |
| C6H12N2Na2O5S2 | |
| MFCD00243291 | |
| disodium l-cystine anion hydrate, l-cystine disodium salt monohydrate, h-cys-oh 2.2na.h2o, c6h10n2o4s2.2na.h2o | |
| disodium;(2R)-2-amino-3-[[(2R)-2-amino-2-carboxylatoethyl]disulfanyl]propanoate;hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 265-025-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only