missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Cysteine Hydrochloride Monohydrate, 98.5 to 101.5% (Dry Basis)
CAS: 7048-04-6 | C3H10ClNO3S | 175.63 g/mol
$87.25 - $2568.11
Chemical Identifiers
| CAS | 4-6-7048 |
|---|---|
| Molecular Formula | C3H10ClNO3S |
| Molecular Weight (g/mol) | 175.63 |
| MDL Number | MFCD00065606 |
| InChI Key | QIJRTFXNRTXDIP-JIZZDEOASA-N |
| Synonym | l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.H2O, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl |
| PubChem CID | 23462 |
| IUPAC Name | (2 R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride |
| SMILES | C(C(C(=O)O)N)S.O.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AAJ1403514
|
Thermo Fisher Scientific
J1403514 |
25 g | Plastic Bottle |
Each for $87.25
|
|
||||
|
AAJ1403522
|
Thermo Fisher Scientific
J1403522 |
100 g | Plastic Bottle |
Each for $141.25
|
|
||||
|
AAJ14035A1
|
Thermo Fisher Scientific
J14035A1 |
1 kg | Plastic Bottle |
Each for $594.38
|
|
||||
|
AAJ14035A7
|
Thermo Fisher Scientific
J14035A7 |
5 kg | Plastic Bottle |
Each for $2,568.11
|
|
||||
Chemical Identifiers
| 4-6-7048 | |
| 175.63 | |
| QIJRTFXNRTXDIP-JIZZDEOASA-N | |
| 23462 | |
| C(C(C(=O)O)N)S.O.Cl |
| C3H10ClNO3S | |
| MFCD00065606 | |
| l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.H2O, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl | |
| (2 R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride |
Specifications
| ∼175°C (decomposition) | |
| C3H10ClNO3S | |
| HSCH2CH(NH2)COOH.HCl.H2O | |
| 5158059 | |
| QIJRTFXNRTXDIP-JIZZDEOASA-N | |
| (2 R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride | |
| 25 g | |
| 175.63 | |
| Crystalline Powder | |
| Dry basis |
| 4-6-7048 | |
| Plastic Bottle | |
| MFCD00065606 | |
| l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.H2O, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl | |
| C(C(C(=O)O)N)S.O.Cl | |
| 175.63 | |
| 23462 | |
| 98.5 to 101.5% | |
| L-Cysteine Hydrochloride Monohydrate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-157-7
RTECSNumber : HA2285000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only