missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine Hydrochloride Monohydrate, 98.5 to 101.5% (Dry Basis)

CAS: 7048-04-6 | C3H10ClNO3S | 175.63 g/mol

Supplier:  Thermo Fisher Scientific J14035A1

 View more versions of this product

Catalog No. AAJ14035A1


Only null left

Description

Description

Specifications

Specifications

∼175°C (decomposition)
4-6-7048
C3H10ClNO3S
HSCH2CH(NH2)COOH.HCl.H2O
5158059
QIJRTFXNRTXDIP-JIZZDEOASA-N
(2 R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
1 kg
175.63
Crystalline Powder
L-Cysteine Hydrochloride Monohydrate
Dry basis
Plastic Bottle
MFCD00065606
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.H2O, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
C(C(C(=O)O)N)S.O.Cl
175.63
23462
98.5 to 101.5%

Chemical Identifiers

4-6-7048
175.63
QIJRTFXNRTXDIP-JIZZDEOASA-N
23462
C(C(C(=O)O)N)S.O.Cl
C3H10ClNO3S
MFCD00065606
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.H2O, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
(2 R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-157-7

RTECSNumber : HA2285000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only