missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ L-Cysteine, Hydrochloride, >98%, Calbiochem™,
$128.07 - $572.00
Chemical Identifiers
| CAS | 52-89-1 |
|---|---|
| Molecular Formula | C3H8ClNO2S |
| Molecular Weight (g/mol) | 157.61 |
| MDL Number | MFCD00064553 |
| InChI Key | IFQSXNOEEPCSLW-QYLAHFPANA-N |
| Synonym | l-cysteine hydrochloride, cystein chloride, r-cysteine hydrochloride, l-cysteine.hcl, cysteine monohydrochloride, l-+-cysteine hydrochloride, l-cysteine, hydrochloride, l-cysteine monohydrochloride, cysteine hcl, l-cysteine hydrochloride anhydrous |
| PubChem CID | 60960 |
| IUPAC Name | (2R)-2-amino-3-sulfanylpropanoic acid hydrochloride |
| SMILES | Cl.N[C@@H](CS)C(O)=O |
Chemical Identifiers
| C3H8ClNO2S | |
| MFCD00064553 | |
| l-cysteine hydrochloride, cystein chloride, r-cysteine hydrochloride, l-cysteine.hcl, cysteine monohydrochloride, l-+-cysteine hydrochloride, l-cysteine, hydrochloride, l-cysteine monohydrochloride, cysteine hcl, l-cysteine hydrochloride anhydrous | |
| (2R)-2-amino-3-sulfanylpropanoic acid hydrochloride |
Specifications
| 52-89-1 | |
| C3H8ClNO2S | |
| l-cysteine hydrochloride, cystein chloride, r-cysteine hydrochloride, l-cysteine.hcl, cysteine monohydrochloride, l-+-cysteine hydrochloride, l-cysteine, hydrochloride, l-cysteine monohydrochloride, cysteine hcl, l-cysteine hydrochloride anhydrous | |
| Cl.N[C@@H](CS)C(O)=O | |
| 157.61 | |
| >98% (Titration) |
| White | |
| MFCD00064553 | |
| IFQSXNOEEPCSLW-QYLAHFPANA-N | |
| (2R)-2-amino-3-sulfanylpropanoic acid hydrochloride | |
| 60960 | |
| Solid |