missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ L-Cysteine, Hydrochloride, >98%, Calbiochem™,

Catalog No. 2430100GM
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2430100GM 100 g Glass Bottle
24301KG 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2430100GM Supplier MilliporeSigma™ Supplier No. 2430100GM

Chemical Identifiers

CAS 52-89-1
Molecular Formula C3H8ClNO2S
Molecular Weight (g/mol) 157.61
MDL Number MFCD00064553
InChI Key IFQSXNOEEPCSLW-QYLAHFPANA-N
Synonym l-cysteine hydrochloride, cystein chloride, r-cysteine hydrochloride, l-cysteine.hcl, cysteine monohydrochloride, l-+-cysteine hydrochloride, l-cysteine, hydrochloride, l-cysteine monohydrochloride, cysteine hcl, l-cysteine hydrochloride anhydrous
PubChem CID 60960
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid hydrochloride
SMILES Cl.N[C@@H](CS)C(O)=O

Specifications

Color White
Packaging Glass Bottle
Quantity 100 g
Percent Purity >98% (Titration)
Physical Form Solid