missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ L-Cysteine, Hydrochloride, >98%, Calbiochem™,

Supplier:  MilliporeSigma™ 24301KG

 View more versions of this product

Catalog No. 24301KG


Only null left

Description

Description

Specifications

Specifications

52-89-1
C3H8ClNO2S
MFCD00064553
IFQSXNOEEPCSLW-QYLAHFPANA-N
(2R)-2-amino-3-sulfanylpropanoic acid hydrochloride
157.61
>98% (Titration)
White
Glass Bottle
l-cysteine hydrochloride, cystein chloride, r-cysteine hydrochloride, l-cysteine.hcl, cysteine monohydrochloride, l-+-cysteine hydrochloride, l-cysteine, hydrochloride, l-cysteine monohydrochloride, cysteine hcl, l-cysteine hydrochloride anhydrous
Cl.N[C@@H](CS)C(O)=O
1 kg
60960
Solid

Chemical Identifiers

52-89-1
157.61
IFQSXNOEEPCSLW-QYLAHFPANA-N
60960
Cl.N[C@@H](CS)C(O)=O
C3H8ClNO2S
MFCD00064553
l-cysteine hydrochloride, cystein chloride, r-cysteine hydrochloride, l-cysteine.hcl, cysteine monohydrochloride, l-+-cysteine hydrochloride, l-cysteine, hydrochloride, l-cysteine monohydrochloride, cysteine hcl, l-cysteine hydrochloride anhydrous
(2R)-2-amino-3-sulfanylpropanoic acid hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions