missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ L-Cysteine, Free Base, ≥98%, Calbiochem™,
$343.00 - $956.00
Chemical Identifiers
| CAS | 52-90-4 |
|---|---|
| Molecular Formula | C3H7NO2S |
| Molecular Weight (g/mol) | 121.154 |
| InChI Key | XUJNEKJLAYXESH-REOHCLBHSA-N |
| Synonym | l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid |
| PubChem CID | 5862 |
| ChEBI | CHEBI:17561 |
| IUPAC Name | (2R)-2-amino-3-sulfanylpropanoic acid |
| SMILES | C(C(C(=O)O)N)S |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
243005100GM
|
MilliporeSigma™
243005100GM |
100 g | Glass Bottle |
Each for $343.00
|
|
||||
|
2430051KG
|
MilliporeSigma™
2430051KG |
1 kg | Glass Bottle |
Each for $956.00
|
|
||||
Chemical Identifiers
| 52-90-4 | |
| 121.154 | |
| l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid | |
| CHEBI:17561 | |
| C(C(C(=O)O)N)S |
| C3H7NO2S | |
| XUJNEKJLAYXESH-REOHCLBHSA-N | |
| 5862 | |
| (2R)-2-amino-3-sulfanylpropanoic acid |
Specifications
| 52-90-4 | |
| C3H7NO2S | |
| XUJNEKJLAYXESH-REOHCLBHSA-N | |
| (2R)-2-amino-3-sulfanylpropanoic acid | |
| 5862 | |
| ≥98% (Perchloric Acid Titration) |
| White | |
| l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid | |
| C(C(C(=O)O)N)S | |
| 121.154 | |
| CHEBI:17561 | |
| Solid |