missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ L-Cysteine, Free Base, ≥98%, Calbiochem™,

Catalog No. 243005100GM
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
243005100GM 100 g Glass Bottle
2430051KG 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 243005100GM Supplier MilliporeSigma™ Supplier No. 243005100GM

Chemical Identifiers

CAS 52-90-4
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.154
InChI Key XUJNEKJLAYXESH-REOHCLBHSA-N
Synonym l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
PubChem CID 5862
ChEBI CHEBI:17561
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid
SMILES C(C(C(=O)O)N)S

Specifications

Color White
Packaging Glass Bottle
Quantity 100 g
Percent Purity ≥98% (Perchloric Acid Titration)
Physical Form Solid