missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ L-Cysteine, Free Base, ≥98%, Calbiochem™,

Supplier:  MilliporeSigma™ 2430051KG

 View more versions of this product

Catalog No. 2430051KG


Only null left

Description

Description

Specifications

Specifications

52-90-4
C3H7NO2S
l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
C(C(C(=O)O)N)S
1 kg
5862
≥98% (Perchloric Acid Titration)
White
Glass Bottle
XUJNEKJLAYXESH-REOHCLBHSA-N
(2R)-2-amino-3-sulfanylpropanoic acid
121.154
CHEBI:17561
Solid

Chemical Identifiers

52-90-4
121.154
l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
CHEBI:17561
C(C(C(=O)O)N)S
C3H7NO2S
XUJNEKJLAYXESH-REOHCLBHSA-N
5862
(2R)-2-amino-3-sulfanylpropanoic acid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions