missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteic Acid 98.0+%, TCI America™

$36.57 - $248.34

Chemical Identifiers

CAS 498-40-8
Molecular Formula C3H7NO5S
Molecular Weight (g/mol) 169.15
MDL Number MFCD00007524
InChI Key XVOYSCVBGLVSOL-UHFFFAOYNA-N
Synonym l-cysteic acid, cysteinesulfonic acid, l-alanine, 3-sulfo, 3-sulfo-l-alanine, cysteic acid, l, r-2-amino-3-sulfopropanoic acid, cysteic acid van, l-cysteate, cysteinic acid, cysteric acid
PubChem CID 72886
ChEBI CHEBI:17285
IUPAC Name 2-amino-3-sulfopropanoic acid
SMILES NC(CS(O)(=O)=O)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C05141G
View Documents
TCI America
C05141G
1 g
Each for $36.57
Only null left
 
C051425G
missing translation for 'viewDocuments'
TCI America
C051425G
25 g
Each for $248.34
Only null left
 
Description

Description

Specifications

Chemical Identifiers

498-40-8
169.15
XVOYSCVBGLVSOL-UHFFFAOYNA-N
72886
2-amino-3-sulfopropanoic acid
C3H7NO5S
MFCD00007524
l-cysteic acid, cysteinesulfonic acid, l-alanine, 3-sulfo, 3-sulfo-l-alanine, cysteic acid, l, r-2-amino-3-sulfopropanoic acid, cysteic acid van, l-cysteate, cysteinic acid, cysteric acid
CHEBI:17285
NC(CS(O)(=O)=O)C(O)=O

Specifications

498-40-8
C3H7NO5S
1 g
XVOYSCVBGLVSOL-UHFFFAOYNA-N
2-amino-3-sulfopropanoic acid
72886
169.15
Crystalline Powder
White
MFCD00007524
l-cysteic acid, cysteinesulfonic acid, l-alanine, 3-sulfo, 3-sulfo-l-alanine, cysteic acid, l, r-2-amino-3-sulfopropanoic acid, cysteic acid van, l-cysteate, cysteinic acid, cysteric acid
NC(CS(O)(=O)=O)C(O)=O
169.15
CHEBI:17285
≥98.0% (T)
L-Cysteic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents