accessibility menu, dialog, popup

UserName

NH2SOHOOOHO

CAS RN 498-40-8

IUPAC Name: 2-amino-3-sulfopropanoic acid
Synonyms: cysteic acid acid, cysteic cysteate cysteinesulfonate β-sulfoalanine cysteine sulfonic acid 3-sulfoalanine
Molecular Weight (g/mol): 169.15
Molecular Formula: C3H7NO5S
InChi Key: XVOYSCVBGLVSOL-UHFFFAOYNA-N
SMILES: NC(CS(O)(=O)=O)C(O)=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)