missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteic Acid 98.0+%, TCI America™

Catalog No. C051425G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C051425G 25 g
C05141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C051425G Supplier TCI America Supplier No. C051425G
Only null left

Chemical Identifiers

CAS 498-40-8
Molecular Formula C3H7NO5S
Molecular Weight (g/mol) 169.15
MDL Number MFCD00007524
InChI Key XVOYSCVBGLVSOL-UHFFFAOYNA-N
Synonym l-cysteic acid, cysteinesulfonic acid, l-alanine, 3-sulfo, 3-sulfo-l-alanine, cysteic acid, l, r-2-amino-3-sulfopropanoic acid, cysteic acid van, l-cysteate, cysteinic acid, cysteric acid
PubChem CID 72886
ChEBI CHEBI:17285
IUPAC Name 2-amino-3-sulfopropanoic acid
SMILES NC(CS(O)(=O)=O)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 169.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Cysteic Acid