missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteic acid monohydrate, 99%, Thermo Scientific Chemicals

Catalog No. p-7077396
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452840100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC452840100 Supplier Thermo Scientific Chemicals Supplier No. 452840100
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23537-25-9
Molecular Formula C3H6NO5S
Molecular Weight (g/mol) 168.14
MDL Number MFCD00149544
InChI Key XVOYSCVBGLVSOL-REOHCLBHSA-M
Synonym l-cysteic acid monohydrate, unii-vn8x20t18r, cysteic acid monohydrate, l, r-2-amino-3-sulfopropanoic acid hydrate, l-alanine, 3-sulfo-, monohydrate, cysteinesulfonic acid hydrate, h-cys o 2-oh.h2o, l-cysteic acid hydrate, l-cysteic acid monohydrate mi, alanine, 3-sulfo-, monohydrate, l
PubChem CID 12308854
IUPAC Name (2R)-2-amino-3-sulfopropanoic acid;hydrate
SMILES [NH3+][C@@H](CS([O-])(=O)=O)C([O-])=O

Specifications

Melting Point 253.0°C to 254.5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Linear Formula HO3SCH2CH(NH2)CO2H·H2O
Quantity 10 g
Specific Rotation Condition + 8.00 (20.00°C c=5, H2O)
Specific Rotation + 8.00
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 187.17
Percent Purity 99%
Chemical Name or Material L-Cysteic acid monohydrate
Show More Show Less

RUO – Research Use Only