missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteic acid, 99%

Catalog No. AC226120100
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC226120100 10 g
AC226120500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226120100 Supplier Thermo Scientific Chemicals Supplier No. 226120100

CAS: 498-40-8 | C3H7NO5S | 169.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 498-40-8
Molecular Formula C3H7NO5S
Molecular Weight (g/mol) 169.15
MDL Number MFCD00007524
InChI Key XVOYSCVBGLVSOL-UHFFFAOYNA-N
Synonym l-cysteic acid, cysteinesulfonic acid, l-alanine, 3-sulfo, 3-sulfo-l-alanine, cysteic acid, l, r-2-amino-3-sulfopropanoic acid, cysteic acid van, l-cysteate, cysteinic acid, cysteric acid
PubChem CID 72886
ChEBI CHEBI:17285
SMILES NC(CS(O)(=O)=O)C(O)=O

Specifications

Melting Point 267.0°C to 269.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HO3SCH2CH(NH2)CO2H
Quantity 10 g
Beilstein 04,533
Merck Index 15,2777
Specific Rotation 8.3
Solubility Information (5% in water) Clear colorless
IUPAC Name (2R)-2-amino-3-sulfopropanoic acid
Formula Weight 169.16
Percent Purity 99%
Physical Form Crystalline Powder or Powder
Chemical Name or Material L-Cysteic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 207-861-3

RUO – Research Use Only