missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Citrulline, 99.3%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210139780
| Quantity | 100 g |
|---|
Chemical Identifiers
| C6H13N3O3 | |
| MFCD00064397 | |
| l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l | |
| CHEBI:16349 |
Specifications
| L-Citrulline | |
| White | |
| C6H13N3O3 | |
| l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l | |
| NC(CCCNC(N)=O)C(O)=O | |
| 100 g | |
| 9750 | |
| 175.19 |
| 372-75-8 | |
| 99.3% | |
| MFCD00064397 | |
| RHGKLRLOHDJJDR-UHFFFAOYNA-N | |
| (2S)-2-amino-5-(carbamoylamino)pentanoic acid | |
| 175.19 | |
| CHEBI:16349 | |
| Crystalline Powder |