missing translation for 'onlineSavingsMsg'
Learn More

L-Citrulline, 99.3%, MP Biomedicals™

Catalog No. ICN10139725
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10139725 25 g
ICN10139780 100 g
ICN10139790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10139725 Supplier MP Biomedicals Inc Supplier No. 0210139725

Chemical Identifiers

CAS 372-75-8
Molecular Formula C6H13N3O3
Molecular Weight (g/mol) 175.19
MDL Number MFCD00064397
InChI Key RHGKLRLOHDJJDR-UHFFFAOYNA-N
Synonym l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l
PubChem CID 9750
ChEBI CHEBI:16349
SMILES NC(CCCNC(N)=O)C(O)=O

Specifications

Color White
Assay Percent Range 99.3%
IUPAC Name 2-amino-5-(carbamoylamino)pentanoic acid
Quantity 25 g
Formula Weight 175.19
Physical Form Crystalline Powder
Chemical Name or Material L-Citrulline