missing translation for 'onlineSavingsMsg'
Learn More

L-Citrulline, 98%

Catalog No. p-7021114
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1331609 10 g
AAA1331618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1331609 Supplier Thermo Scientific Chemicals Supplier No. A1331609
Only null left

CAS: 372-75-8 | C6H13N3O3 | 175.19 g/mol

L-Citrulline, is used as an essential intermediate in the biosynthesis of nitric oxide from L-arginine . It is also used as a nutritional drink and biochemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Citrulline, is used as an essential intermediate in the biosynthesis of nitric oxide from L-arginine . It is also used as a nutritional drink and biochemical reagent.

Solubility
It is soluble in water.

Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.

Chemical Identifiers

CAS 372-75-8
Molecular Formula C6H13N3O3
Molecular Weight (g/mol) 175.19
MDL Number MFCD00064397
InChI Key RHGKLRLOHDJJDR-UHFFFAOYNA-N
Synonym l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l
PubChem CID 9750
ChEBI CHEBI:16349
IUPAC Name (2S)-2-amino-5-(carbamoylamino)pentanoic acid
SMILES NC(CCCNC(N)=O)C(O)=O

Specifications

Melting Point 212°C to 214°C (decomposition)
Quantity 10 g
Beilstein 6055157
Merck Index 14,2331
Solubility Information It is soluble in water.
Optical Rotation +22° (c=2 in 1N HCl)
Formula Weight 175.19
Percent Purity 98%
Chemical Name or Material L-Citrulline
EINECSNumber 206-759-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only