missing translation for 'onlineSavingsMsg'
Learn More

L-Carnitine-(methyl-d3) inner salt, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012858
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012858 1 mg
111012860 5 mg
111012859 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111012858 Supplier Merck Emd Millipore Supplier No. 949911MG
Only null left

Specifications

CAS 126372-70-1
Linear Formula C7H12D3 NO3
Synonym (R)-3-Carboxy-2-hydroxy-N,N-dimethyl-N-(methyl-d3)-1-propanaminium inner salt
Type Analytical Standard
Quantity 1 mg
Formula Weight 164.22
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard