missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Carnitine L-Tartrate 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 236.24 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-Carnitine L-Tartrate |
Safety and Handling
| TSCA | No |