accessibility menu, dialog, popup

UserName

N(R)H3CH3CH3CHOOOHHO(R)(R)OOOHOHOH

CAS RN 36687-82-8

IUPAC Name: (2R,3R)-2,3-dihydroxybutanedioic acid (3R)-3-hydroxy-4-(trimethylazaniumyl)butanoate
Synonyms: L(+)-tartaric acid; L-carnitine
Molecular Weight (g/mol): 311.29
Molecular Formula: C11H21NO9
InChi Key: RZALONVQKUWRRY-HMWYIXBMNA-N
SMILES: O[C@H]([C@@H](O)C(O)=O)C(O)=O.[H][C@@](O)(CC([O-])=O)C[N+](C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results