missing translation for 'onlineSavingsMsg'
Learn More

L-Canavanine Sulfate Hydrate 97.0+%, TCI America™

Catalog No. c0018100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0018100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C0018100MG Supplier TCI America Supplier No. C0018100MG

Chemical Identifiers

CAS 2219-31-0
Molecular Formula C5H14N4O7S
Molecular Weight (g/mol) 274.248
MDL Number MFCD00012618
InChI Key MVIPJKVMOKFIEV-UHFFFAOYSA-N
Synonym l-canavanine sulfate, canavanine sulfate, canavanine sulphate, unii-pn00q697sg, l-canavanine sulfate salt, l-canavanine sulphate, s-2-amino-4-guanidinooxy butanoic acid compound with sulfuric acid 1:1, l-2-amino-4-guanidiooxy butyric acid sulfate, canavanine; sulfuric acid, canavanine monosulfate
PubChem CID 24212197
IUPAC Name 2-amino-4-(diaminomethylideneamino)oxybutanoic acid;sulfuric acid
SMILES C(CON=C(N)N)C(C(=O)O)N.OS(=O)(=O)O

Specifications

Melting Point 160°C
Color White
Quantity 100 mg
Formula Weight 274.25
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Canavanine Sulfate Hydrate