accessibility menu, dialog, popup

UserName

CAS RN 2219-31-0

NH2OHNH2NNH2OHOOHSOOO

CAS RN 2219-31-0

IUPAC Name: {amino[(3-amino-3-carboxypropoxy)amino]methylidene}azanium hydrogen sulfate
Synonyms: {amino[(3-amino-3-carboxypropoxy)amino]methylidene}azanium hydrogen sulfate
Molecular Weight (g/mol): 274.25
Molecular Formula: C5H14N4O7S
InChi Key: MVIPJKVMOKFIEV-UHFFFAOYNA-N
SMILES: OS([O-])(=O)=O.NC(CCONC(N)=[NH2+])C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results