missing translation for 'onlineSavingsMsg'
Learn More

L-Bornyl acetate, 95%

CAS: 5655-61-8 | C12H20O2 | 196.29 g/mol

$50.83 - $105.47

Chemical Identifiers

CAS 5655-61-8
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00867808,MFCD00135942,MFCD00135942
InChI Key KGEKLUUHTZCSIP-HOSYDEDBSA-N
Synonym bornyl acetate, -bornyl acetate
PubChem CID 44630108
IUPAC Name [(1R,3S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate
SMILES CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC106550050
SDS View Documents
Thermo Scientific Chemicals
106550050
5 g
Each for $50.83
Only null left
 
AC106551000
SDS View Documents
Thermo Scientific Chemicals
106551000
100 g
Each for $105.47
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5655-61-8
196.29
KGEKLUUHTZCSIP-HOSYDEDBSA-N
44630108
CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C
C12H20O2
MFCD00867808,MFCD00135942,MFCD00135942
bornyl acetate, -bornyl acetate
[(1R,3S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate

Specifications

5655-61-8
0.9875g/mL
84°C
98% min. (as C12H20O2) Total esters (GC)
C12H20O2
MFCD00867808,MFCD00135942,MFCD00135942
06,82
15,1341
bornyl acetate, -bornyl acetate
KGEKLUUHTZCSIP-HOSYDEDBSA-N
[(1R,3S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate
44630108
95%
L-Bornyl acetate, 95%
Colorless
223°C to 224°C
Authentic
Glass bottle
1.4620 to 1.465
5 g
0.9875
-38
Solubility in water: .. Other solubilities: miscible with 95% alcohol and ether
CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C
196.29
196.29
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-101-4

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only