missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Aspartyl-L-phenylalanine, 95%

CAS: 13433-09-5 | C13H16N2O5 | 280.28 g/mol

$143.61 - $550.60

Chemical Identifiers

CAS 13433-09-5
Molecular Formula C13H16N2O5
Molecular Weight (g/mol) 280.28
MDL Number MFCD00063155
InChI Key YZQCXOFQZKCETR-UWVGGRQHSA-N
Synonym l-aspartyl-l-phenylalanine, aspartyl-phenylalanine, demethylaspartame, aspartylphenylalanine, asp-phe, h-asp-phe-oh, l-a-aspartyl-l-phenylalanine, l-alpha-aspartyl-l-phenylalanine, alpha-aspartylphenylalanine, s-3-amino-4-s-1-carboxy-2-phenylethyl amino-4-oxobutanoic acid
PubChem CID 93078
ChEBI CHEBI:73830
SMILES N[C@@H](CC(O)=O)C(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC310021000
SDS View Documents
Thermo Scientific Chemicals
310021000
100 mg Glass Bottle
Each for $143.61
Only null left
 
AC310025000
SDS View Documents
Thermo Scientific Chemicals
310025000
500 mg Glass Bottle
Each for $550.60
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13433-09-5
280.28
YZQCXOFQZKCETR-UWVGGRQHSA-N
93078
N[C@@H](CC(O)=O)C(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
C13H16N2O5
MFCD00063155
l-aspartyl-l-phenylalanine, aspartyl-phenylalanine, demethylaspartame, aspartylphenylalanine, asp-phe, h-asp-phe-oh, l-a-aspartyl-l-phenylalanine, l-alpha-aspartyl-l-phenylalanine, alpha-aspartylphenylalanine, s-3-amino-4-s-1-carboxy-2-phenylethyl amino-4-oxobutanoic acid
CHEBI:73830

Specifications

13433-09-5
Authentic
C13H16N2O5
HO2CCH2CH(NH2)CONHCH(CH2C6H5)CO2H
+10.0° to +13.0° (24°C, 589nm) (c=1, 0.5N HCl)
3% max. (K.F.)
N[C@@H](CC(O)=O)C(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
100 mg
93078
280.28
Powder
Tan to White
95%
Glass Bottle
MFCD00063155
l-aspartyl-l-phenylalanine, aspartyl-phenylalanine, demethylaspartame, aspartylphenylalanine, asp-phe, h-asp-phe-oh, l-a-aspartyl-l-phenylalanine, l-alpha-aspartyl-l-phenylalanine, alpha-aspartylphenylalanine, s-3-amino-4-s-1-carboxy-2-phenylethyl amino-4-oxobutanoic acid
YZQCXOFQZKCETR-UWVGGRQHSA-N
(3S)-3-amino-3-{[(1S)-1-carboxy-2-phenylethyl]carbamoyl}propanoic acid
280.28
CHEBI:73830
≥94% (HPLC)
L-Aspartyl-L-phenylalanine, 95%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 236-557-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only