accessibility menu, dialog, popup

UserName

NH2(S)OHOONH(S)HOO

CAS RN 13433-09-5

IUPAC Name: (3S)-3-amino-3-{[(1S)-1-carboxy-2-phenylethyl]carbamoyl}propanoic acid
Synonyms: L-aspartyl-L-phenylalanine
Molecular Weight (g/mol): 280.28
Molecular Formula: C13H16N2O5
InChi Key: YZQCXOFQZKCETR-UWVGGRQHSA-N
SMILES: N[C@@H](CC(O)=O)C(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)