missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Aspartyl-L-phenylalanine, 95%

Catalog No. AC310021000
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC310021000 100 mg Glass Bottle
AC310025000 500 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC310021000 Supplier Thermo Scientific Chemicals Supplier No. 310021000
Only null left

CAS: 13433-09-5 | C13H16N2O5 | 280.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13433-09-5
Molecular Formula C13H16N2O5
Molecular Weight (g/mol) 280.28
MDL Number MFCD00063155
InChI Key YZQCXOFQZKCETR-UWVGGRQHSA-N
Synonym l-aspartyl-l-phenylalanine, aspartyl-phenylalanine, demethylaspartame, aspartylphenylalanine, asp-phe, h-asp-phe-oh, l-a-aspartyl-l-phenylalanine, l-alpha-aspartyl-l-phenylalanine, alpha-aspartylphenylalanine, s-3-amino-4-s-1-carboxy-2-phenylethyl amino-4-oxobutanoic acid
PubChem CID 93078
ChEBI CHEBI:73830
SMILES N[C@@H](CC(O)=O)C(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Linear Formula HO2CCH2CH(NH2)CONHCH(CH2C6H5)CO2H
Specific Rotation +10.0° to +13.0° (24°C, 589nm) (c=1, 0.5N HCl)
Water 3% max. (K.F.)
IUPAC Name (3S)-3-amino-3-{[(1S)-1-carboxy-2-phenylethyl]carbamoyl}propanoic acid
Quantity 100 mg
Formula Weight 280.28
Percent Purity ≥94% (HPLC)
Physical Form Powder
Chemical Name or Material L-Aspartyl-L-phenylalanine, 95%
Show More Show Less
EINECSNumber 236-557-3

RUO – Research Use Only