missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic Acid Sodium Salt, Monohydrate, 98-101%, Spectrum™ Chemical

Catalog No. p-7156944
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603729 25 g Poly Bottle
18603730 100 g Poly Bottle
18603731 1 kg Poly Bottle
18603733 12 kg Poly Pail
18603732 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18603729 Supplier Spectrum Chemical Mfg Cor Supplier No. A137525GM
Only null left

A1375, 323194-76-9, C4H6NO4Na·H2O

Spectrum™ Chemical L-Aspartic Acid Sodium Salt, Monohydrate

Chemical Identifiers

CAS 323194-76-9
Molecular Formula C4H8NNaO5
Molecular Weight (g/mol) 173.10
MDL Number MFCD00152960
InChI Key PPTHNBYUFXSJPS-UHFFFAOYNA-M
IUPAC Name sodium 2-amino-3-carboxypropanoate hydrate
SMILES O.[Na+].NC(CC(O)=O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 25 g
Percent Purity 98 to 101%
Grade Reagent
Loss on Drying 0.3%
Packaging Poly Bottle