accessibility menu, dialog, popup

UserName

CAS RN 323194-76-9

IUPAC Name: sodium 2-amino-3-carboxypropanoate hydrate
Synonyms: sodium 2-amino-3-carboxypropanoate hydrate
Molecular Weight (g/mol): 173.1
Molecular Formula: C4H8NNaO5
InChi Key: PPTHNBYUFXSJPS-UHFFFAOYNA-M
SMILES: O.[Na+].NC(CC(O)=O)C([O-])=O
Percent Purity 
  • (5)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)