missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014660
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014660 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014660 Supplier Merck Emd Millipore Supplier No. PHR11041G
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 56-84-8
Physical Form Neat
Linear Formula HO2 CCH2 CH(NH2)CO2 H
MDL Number MFCD00002616
Melting Point >300°C
Synonym (S)-(+)-Aminosuccinic acid; (S)-Aminobutanedioic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 133.10
Grade Certified Reference Material