missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Aspartic acid monosodium salt monohydrate, 99%

Catalog No. AAB2232122
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2232122 100 g
AAB2232136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2232122 Supplier Thermo Scientific Chemicals Supplier No. B2232122

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 323194-76-9 | C4H8NNaO5 | 173.10 g/mol

L-Aspartic acid monosodium salt monohydrate is used as a principal neurotransmitter for fast synaptic excitation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid monosodium salt monohydrate is used as a principal neurotransmitter for fast synaptic excitation.

Solubility
Soluble in cold water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 323194-76-9
Molecular Formula C4H8NNaO5
Molecular Weight (g/mol) 173.10
MDL Number MFCD00152960
InChI Key PPTHNBYUFXSJPS-UHFFFAOYNA-M
Synonym sodium l-aspartate, l-aspartic acid sodium salt monohydrate, sodium aspartate monohydrate, sodium l-aspartate monohydrate, s-2-aminobutanedioic acid sodium salt, l-aspartic acid monosodium salt monohydrate, l-aspartic acid, monosodium salt, monohydrate, sodium +-aspartate hydrate, l-aspartic acid sodium salt monohydrate tlc, l-aspartic acid sodium salt monohydrate nt
PubChem CID 23679051
SMILES O.[Na+].NC(CC(O)=O)C([O-])=O

Specifications

Solubility Information Soluble in cold water.
IUPAC Name sodium 2-amino-3-carboxypropanoate hydrate
Quantity 100 g
Formula Weight 173.11 (155.10 Anhydrous)
Percent Purity 99%
Chemical Name or Material L-Aspartic acid monosodium salt monohydrate
EINECSNumber 223-264-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only