missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Aspartic acid dimethyl ester hydrochloride, 98%

Catalog No. p-7043171
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6598306 5 g
AAJ6598314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6598306 Supplier Thermo Scientific Chemicals Supplier No. J6598306

missing translation for 'validMainframeMsg'

CAS: 32213-95-9 | C6H12ClNO4 | 197.615 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (almost transparency) and methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 32213-95-9
Molecular Formula C6H12ClNO4
Molecular Weight (g/mol) 197.615
MDL Number MFCD00038878
InChI Key PNLXWGDXZOYUKB-WCCKRBBISA-N
Synonym h-asp ome-ome hcl, l-aspartic acid dimethyl ester hydrochloride, dimethyl l-aspartate hydrochloride, h-asp ome-ome.hcl, methyl aspartic acid hydrochloride, s-dimethyl 2-aminosuccinate hydrochloride, aspartic acid dimethyl ester hydrochloride, l-aspartic acid dimethyl ester hydro-chloride, s-aminosuccinic acid dimethyl ester hydrochloride, dimethyl aspartic acid hydrochloride
PubChem CID 2734892
IUPAC Name dimethyl (2S)-2-aminobutanedioate;hydrochloride
SMILES COC(=O)CC(C(=O)OC)N.Cl

Specifications

Melting Point 115°C to 120°C
Color White
Solubility Information Soluble in water (almost transparency) and methanol.
Quantity 5 g
Formula Weight 197.62
Optical Rotation +15° (c=1 in Methanol)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Aspartic acid dimethyl ester hydrochloride
EINECSNumber 250-957-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only