missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Aspartic acid, 98+%
CAS: 56-84-8 | C4H7NO4 | 133.10 g/mol
$48.04 - $654.60
Chemical Identifiers
| CAS | 56-84-8 |
|---|---|
| Molecular Formula | C4H7NO4 |
| Molecular Weight (g/mol) | 133.10 |
| MDL Number | MFCD00002616 |
| InChI Key | CKLJMWTZIZZHCS-UHFFFAOYNA-N |
| Synonym | l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid |
| PubChem CID | 5960 |
| ChEBI | CHEBI:17053 |
| SMILES | NC(CC(O)=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1352022
|
Thermo Scientific Chemicals
A1352022 |
100 g |
Each for $48.04
|
|
|||||
|
AAA1352030
|
Thermo Scientific™
A1352030 |
250 g |
Each for $63.90
|
|
|||||
|
AAA135200B
|
Thermo Scientific Chemicals
A135200B |
1000 g |
Each for $172.71
|
|
|||||
|
AAA135200I
|
Thermo Scientific Chemicals
A135200I |
5000 g |
Each for $654.60
|
|
|||||
Description
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.
Solubility
Soluble in water(4.5g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
| 56-84-8 | |
| 133.10 | |
| CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
| 5960 | |
| NC(CC(O)=O)C(O)=O |
| C4H7NO4 | |
| MFCD00002616 | |
| l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid | |
| CHEBI:17053 |
Specifications
| >300°C (decomposition) | |
| C4H7NO4 | |
| 1723530 | |
| l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid | |
| CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
| NC(CC(O)=O)C(O)=O | |
| 133.10 | |
| 5960 | |
| Odorless | |
| 133.1 | |
| L-Aspartic acid |
| 56-84-8 | |
| MFCD00002616 | |
| 14,840 | |
| Soluble in water(4.5g/L). | |
| 1.66 | |
| 2-aminobutanedioic acid | |
| 100 g | |
| CHEBI:17053 | |
| +25° (c=2 in 5N HCl) | |
| ≥98% |
Safety and Handling
EINECSNumber : 200-291-6
RTECSNumber : CI9098500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only