missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Aspartic acid, 98+%

CAS: 56-84-8 | C4H7NO4 | 133.10 g/mol

Supplier:  Thermo Scientific™ A1352030

 View more versions of this product

Catalog No. AAA1352030


Only null left

Description

Description

L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.

Solubility
Soluble in water(4.5g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

>300°C (decomposition)
56-84-8
MFCD00002616
14,840
Soluble in water(4.5g/L).
1.66
(2S)-2-aminobutanedioic acid
250 g
CHEBI:17053
+25° (c=2 in 5N HCl)
≥98%
L-Aspartic acid
C4H7NO4
1723530
l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid
CKLJMWTZIZZHCS-UHFFFAOYNA-N
NC(CC(O)=O)C(O)=O
133.10
5960
Odorless
133.1

Chemical Identifiers

56-84-8
133.10
CKLJMWTZIZZHCS-UHFFFAOYNA-N
5960
NC(CC(O)=O)C(O)=O
C4H7NO4
MFCD00002616
l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid
CHEBI:17053
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-291-6

RTECSNumber : CI9098500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only