missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Aspartic acid, 98+%

CAS: 56-84-8 | C4H7NO4 | 133.10 g/mol

Supplier:  Thermo Scientific Chemicals 105040100

 View more versions of this product

Catalog No. AC105040100


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

>230.0°C
98+%
56-84-8
200ppm max.
Authentic
0.5% max. (105°C, 3 hrs)
0.5% max. (TLC)
Plastic Drum
MFCD00002616
300ppm max.
l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid
CKLJMWTZIZZHCS-UHFFFAOYNA-N
(2S)-2-aminobutanedioic acid
133.10
5960
133.1
Crystalline Powder or Crystals
L(+)-Aspartic acid
200ppm max.
White
10ppm max.
10ppm max.
98+%
C4H7NO4
HO2CCH2CH(NH2)CO2H
+ 25.00
0.1% max.
Solubility in water: 5g/L water (25°C). Other solubilities: 2.1g/L water (0°C), 8.5g/L water (40°C), 28.7g/L water (75°C), 16 mg/100 mL ethanol (25°C), soluble in acids, alkalies
NC(CC(O)=O)C(O)=O
+ 25.00 (20.00°C c=8,6N HCl)
10 kg
CHEBI:17053
≥98%

Chemical Identifiers

56-84-8
133.10
CKLJMWTZIZZHCS-UHFFFAOYNA-N
5960
(2S)-2-aminobutanedioic acid
C4H7NO4
MFCD00002616
l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid
CHEBI:17053
NC(CC(O)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-291-6

RTECSNumber : CI9098500

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only