missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Aspartic acid, 98+%

Catalog No. p-61894
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105041000 100 g Plastic Bottle
AC105045000 500 g Plastic Bottle
AC105040025 2.5 kg Plastic Bottle
AC105040100 10 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC105041000 Supplier Thermo Scientific Chemicals Supplier No. 105041000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56-84-8 | C4H7NO4 | 133.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-84-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00002616
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Synonym l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid
PubChem CID 5960
ChEBI CHEBI:17053
IUPAC Name (2S)-2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

Melting Point >230.0°C
Ammonium 200ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 98+%
Loss on Drying 0.5% max. (105°C, 3 hrs)
Other Amino Acids 0.5% max. (TLC)
Packaging Plastic Bottle
Linear Formula HO2CCH2CH(NH2)CO2H
Specific Rotation + 25.00
Sulfate 300ppm max.
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 5g/L water (25°C). Other solubilities: 2.1g/L water (0°C), 8.5g/L water (40°C), 28.7g/L water (75°C), 16 mg/100 mL ethanol (25°C), soluble in acids, alkalies
Quantity 100 g
Formula Weight 133.1
Percent Purity ≥98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Aspartic acid, 98+%
Show More Show Less
EINECSNumber 200-291-6
RTECSNumber CI9098500
TSCA TSCA

RUO – Research Use Only