missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Aspartic Acid, 98.5%, Spectrum™ Chemical
AS125, 56-84-8, C4H7NO4
$117.50 - $438.52
Chemical Identifiers
| CAS | 56-84-8 |
|---|---|
| Molecular Formula | C4H7NO4 |
| Molecular Weight (g/mol) | 133.10 |
| MDL Number | MFCD00002616 |
| InChI Key | CKLJMWTZIZZHCS-UHFFFAOYNA-N |
| IUPAC Name | 2-aminobutanedioic acid |
| SMILES | NC(CC(O)=O)C(O)=O |
Description
Spectrum™ Chemical L-Aspartic Acid is a nonessential amino acid. Aspartic acid is used as a component of parenteral and enteral nutrition. In pharmaceutical agents aspartic acid is used as an ammoniac detoxicating agent, hepar function accelerator and fatigue refresher. Aspartic acid is also widely utilized for mineral supplementation in the form of its potassium, magnesium or calcium salts. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 56-84-8 | |
| 133.10 | |
| CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
| NC(CC(O)=O)C(O)=O |
| C4H7NO4 | |
| MFCD00002616 | |
| 2-aminobutanedioic acid |
Specifications
| 56-84-8 | |
| 10 ppm | |
| 5 ppm | |
| C4H7NO4 | |
| MFCD00002616 | |
| CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
| 2-aminobutanedioic acid | |
| 133.10 | |
| Reagent |
| 100% | |
| 0.1% | |
| 0.2% | |
| Poly Bottle | |
| +24.5° to +26.0° | |
| NC(CC(O)=O)C(O)=O | |
| 100 g | |
| 98.5% |