missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic Acid, 98.5%, Spectrum™ Chemical

Catalog No. 18603727
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603727 100 g Poly Bottle
18603728 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18603727 Supplier Spectrum Chemical Mfg Cor Supplier No. AS125100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

AS125, 56-84-8, C4H7NO4

Spectrum™ Chemical L-Aspartic Acid is a nonessential amino acid. Aspartic acid is used as a component of parenteral and enteral nutrition. In pharmaceutical agents aspartic acid is used as an ammoniac detoxicating agent, hepar function accelerator and fatigue refresher. Aspartic acid is also widely utilized for mineral supplementation in the form of its potassium, magnesium or calcium salts. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 56-84-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00002616
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
IUPAC Name 2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

CAS Max % 100%
Heavy Metals 10 ppm
Ignition Residue 0.1%
Lead (Pb) 5 ppm
Loss on Drying 0.2%
Packaging Poly Bottle
Specific Rotation +24.5° to +26.0°
Quantity 100 g
Percent Purity 98.5%
Grade Reagent