missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Aspartic acid 4-tert-butyl ester, 98%
CAS: 3057-74-7 | C8H15NO4 | 189.211 g/mol
$173.71 - $527.82
Chemical Identifiers
| CAS | 3057-74-7 |
|---|---|
| Molecular Formula | C8H15NO4 |
| Molecular Weight (g/mol) | 189.211 |
| MDL Number | MFCD00038577 |
| InChI Key | MXWMFBYWXMXRPD-YFKPBYRVSA-N |
| Synonym | h-asp otbu-oh, l-aspartic acid 4-tert-butyl ester, h-asp obut-oh, 4-tert-butyl hydrogen l-aspartate, 2s-2-amino-4-tert-butoxy-4-oxobutanoic acid, s-2-amino-4-tert-butoxy-4-oxobutanoic acid, aspartic acid, 4-tert-butyl ester, asp but, asp obut, asp otbu oh |
| PubChem CID | 7010570 |
| IUPAC Name | (2S)-2-amino-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid |
| SMILES | CC(C)(C)OC(=O)CC(C(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5981003
|
Thermo Scientific Chemicals
H5981003 |
1 g |
Each for $173.71
|
|
|||||
|
AAH5981006
|
Thermo Scientific Chemicals
H5981006 |
5 g |
Each for $527.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3057-74-7 | |
| 189.211 | |
| MXWMFBYWXMXRPD-YFKPBYRVSA-N | |
| 7010570 | |
| CC(C)(C)OC(=O)CC(C(=O)O)N |
| C8H15NO4 | |
| MFCD00038577 | |
| h-asp otbu-oh, l-aspartic acid 4-tert-butyl ester, h-asp obut-oh, 4-tert-butyl hydrogen l-aspartate, 2s-2-amino-4-tert-butoxy-4-oxobutanoic acid, s-2-amino-4-tert-butoxy-4-oxobutanoic acid, aspartic acid, 4-tert-butyl ester, asp but, asp obut, asp otbu oh | |
| (2S)-2-amino-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid |
Specifications
| 3057-74-7 | |
| MFCD00038577 | |
| h-asp otbu-oh, l-aspartic acid 4-tert-butyl ester, h-asp obut-oh, 4-tert-butyl hydrogen l-aspartate, 2s-2-amino-4-tert-butoxy-4-oxobutanoic acid, s-2-amino-4-tert-butoxy-4-oxobutanoic acid, aspartic acid, 4-tert-butyl ester, asp but, asp obut, asp otbu oh | |
| CC(C)(C)OC(=O)CC(C(=O)O)N | |
| 1 g | |
| 7010570 | |
| 189.21 | |
| L-Aspartic acid 4-tert-butyl ester |
| C8H15NO4 | |
| 4671089 | |
| MXWMFBYWXMXRPD-YFKPBYRVSA-N | |
| (2S)-2-amino-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid | |
| 189.211 | |
| −9° (c=2 in Methanol: Water (4:1)) | |
| 98% |
Safety and Handling
EINECSNumber : 221-292-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only